LIF372156737 Life Chemicals Inc.

[1-(3,5-dimethoxybenzyl)-1H-indol-3-yl]acetic acid

  • MDL number MFCD11986884
  • Cas Number : 1105192-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart