LIF372156741 Life Chemicals Inc.

2-{[(4-methoxyphenyl)sulfonyl]amino}-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD11986889
  • Cas Number : 1105192-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart