LIF372156745 Life Chemicals Inc.

1-{2-[(3,5-dimethylphenyl)amino]-2-oxoethyl}piperidine-4-carboxylic acid

  • MDL number MFCD11606219
  • Cas Number : 1082910-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart