LIF372156748 Life Chemicals Inc.

2-[(4-chlorobenzoyl)amino]-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD11564904
  • Cas Number : 1082168-15-7
  • Molecular Weight : 282.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart