LIF372156766 Life Chemicals Inc.

1-(3-methoxyphenyl)guanidine

  • MDL number MFCD04973396
  • Cas Number : 57004-60-1
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart