LIF372156771 Life Chemicals Inc.

1-(3-chlorophenyl)guanidine

  • MDL number MFCD08236718
  • Cas Number : 6145-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart