LIF372156865 Life Chemicals Inc.

5,6-dihydro-1,4-dioxine-2-carbonyl chloride

  • MDL number MFCD13173540
  • Cas Number : 61564-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart