LIF372156888 Life Chemicals Inc.

ethyl 5-bromo-2-chloro-1,3-thiazole-4-carboxylate

  • MDL number MFCD16615260
  • Cas Number : 1125409-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart