LIF372156905 Life Chemicals Inc.

1-(3-chloro-4-methylphenyl)guanidine

  • MDL number MFCD16653179
  • Cas Number : 57004-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart