LIF372156919 Life Chemicals Inc.

N-(3,4,5-trimethoxyphenyl)guanidine

  • MDL number MFCD16653192
  • Cas Number : 57004-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart