LIF372156921 Life Chemicals Inc.

[4-(isopropylthio)phenyl]acetyl chloride

  • MDL number MFCD11986934
  • Cas Number : 1170785-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart