LIF372156936 Life Chemicals Inc.

6-bromo-2-chloro-4-methyl-1,3-benzothiazole

  • MDL number MFCD16653205
  • Cas Number : 1177325-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart