LIF372156949 Life Chemicals Inc.

6-bromo-N,N-dimethyl-1,3-benzothiazol-2-amine

  • MDL number MFCD16653214
  • Cas Number : 75104-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart