LIF372156970 Life Chemicals Inc.

2-[(2-thienylcarbonyl)amino]-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD16631787
  • Cas Number : 1204297-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart