LIF372156976 Life Chemicals Inc.

ethyl 2,4-dimethyl-6-oxo-1,6-dihydropyridine-3-carboxylate

  • MDL number MFCD00514034
  • Cas Number : 36853-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart