LIF372156980 Life Chemicals Inc.

2-hydrazino-6-(trifluoromethoxy)-1,3-benzothiazole

  • MDL number MFCD09749222
  • Cas Number : 133840-98-9
  • Molecular Weight : 249.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart