LIF372156993 Life Chemicals Inc.

4-isopropyl-N-(tetrahydrofuran-2-ylmethyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD16631804
  • Cas Number : 1204297-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart