LIF372157001 Life Chemicals Inc.

5-chloro-4-methyl-N-(3-morpholinopropyl)benzo[d]thiazol-2-amine

  • MDL number MFCD16631817
  • Cas Number : 1204298-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart