LIF372157007 Life Chemicals Inc.

N-(3-(1H-imidazol-1-yl)propyl)-7-chloro-4-methylbenzo[d]thiazol-2-amine

  • MDL number MFCD16631825
  • Cas Number : 1204297-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart