LIF372157009 Life Chemicals Inc.

N,N-diethyl-N'-(6-fluoro-1,3-benzothiazol-2-yl)ethane-1,2-diamine

  • MDL number MFCD16631827
  • Cas Number : 1204297-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart