LIF372157033 Life Chemicals Inc.

2-chloro-8-methyl-4H-pyrido[1,2-a]pyrimidin-4-one

  • MDL number MFCD00857628
  • Cas Number : 17326-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart