LIF372157068 Life Chemicals Inc.

ethyl 1-(2-amino-2-thioxoethyl)piperidine-4-carboxylate

  • MDL number MFCD11052375
  • Cas Number : 1053656-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart