LIF372157092 Life Chemicals Inc.

4-[(4-methyl-1,3-thiazol-2-yl)methoxy]benzoic acid

  • MDL number MFCD09904925
  • Cas Number : 951920-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart