LIF372157148 Life Chemicals Inc.

3-fluorothiophene-2-carboxylic acid

  • MDL number MFCD03426911
  • Cas Number : 32431-84-8
  • Molecular Weight : 146.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart