LIF372157149 Life Chemicals Inc.

methyl 3-fluorothiophene-2-carboxylate

  • MDL number MFCD16606491
  • Cas Number : 100421-52-1
  • Molecular Weight : 160.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart