LIF372157281 Life Chemicals Inc.

1-methanesulfonylazetidine-3-carboxylic acid

  • MDL number MFCD16653314
  • Cas Number : 1219828-27-9
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart