LIF372157283 Life Chemicals Inc.

1-(cyclopropylsulfonyl)azetidine-3-carboxylic acid

  • MDL number MFCD16653316
  • Cas Number : 1219827-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart