LIF372157288 Life Chemicals Inc.

1-isocyanatoadamantane

  • MDL number MFCD00134405
  • Cas Number : 4411-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart