LIF372157291 Life Chemicals Inc.

[3-(prop-2-yn-1-yloxy)phenyl]amine

  • MDL number MFCD12779238
  • Cas Number : 60042-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart