LIF372157295 Life Chemicals Inc.

1-acetylazepan-4-one

  • MDL number MFCD07778600
  • Cas Number : 50492-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart