LIF372157307 Life Chemicals Inc.

N-(4-methylpyrimidin-2-yl)thiourea

  • MDL number MFCD15525460
  • Cas Number : 102739-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart