LIF372157308 Life Chemicals Inc.

N-(5-methylpyridin-2-yl)thiourea

  • MDL number MFCD04117347
  • Cas Number : 131185-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart