LIF372157309 Life Chemicals Inc.

N-(3-methylpyridin-2-yl)thiourea

  • MDL number MFCD03617970
  • Cas Number : 41440-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart