LIF372157322 Life Chemicals Inc.

1-(2-isocyanatoethyl)-4-methoxybenzene

  • MDL number MFCD04974077
  • Cas Number : 52634-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart