LIF372157330 Life Chemicals Inc.

1-(isocyanatomethyl)naphthalene

  • MDL number MFCD11986936
  • Cas Number : 61924-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart