LIF372157334 Life Chemicals Inc.

[5-(4-methoxyphenyl)isoxazol-3-yl]acetic acid

  • MDL number MFCD10036152
  • Cas Number : 1018584-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart