LIF372157338 Life Chemicals Inc.

3-(chloromethyl)-5-(4-fluorophenyl)isoxazole

  • MDL number MFCD06805613
  • Cas Number : 928796-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart