LIF372157341 Life Chemicals Inc.

ethyl 5-amino-1-(4-methoxybenzyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD16631856
  • Cas Number : 384835-92-1
  • Molecular Weight : 275.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart