LIF372157342 Life Chemicals Inc.

7-chloro-4-methyl-1-phenyl-1H-pyrazolo[3,4-d]pyridazine

  • MDL number MFCD16631861
  • Cas Number : 1204297-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart