LIF372157349 Life Chemicals Inc.

2-[5-amino-3-(2-thienyl)-1H-pyrazol-1-yl]-5-ethyl-6-methylpyrimidin-4(3H)-one

  • MDL number MFCD16631874
  • Cas Number : 1207024-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart