LIF372157352 Life Chemicals Inc.

2-[5-amino-3-(2-furyl)-1H-pyrazol-1-yl]-6-isopropylpyrimidin-4(3H)-one

  • MDL number MFCD16631885
  • Cas Number : 1206993-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart