LIF372157356 Life Chemicals Inc.

2-cyclopropylquinazolin-4(3H)-one

  • MDL number MFCD03110630
  • Cas Number : 459796-19-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart