LIF372157363 Life Chemicals Inc.

3-(2-methoxyethyl)-5-methylimidazolidine-2,4-dione

  • MDL number MFCD21091625
  • Cas Number : 1278661-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart