LIF372157386 Life Chemicals Inc.

2-(2,6-Dioxopiperidin-1-yl)ethanesulfonyl chloride

  • MDL number MFCD09864173
  • Cas Number : 1000339-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart