LIF372157395 Life Chemicals Inc.

3-(3-Chloro-4-fluorophenyl)propanoic acid

  • MDL number MFCD04116056
  • Cas Number : 881190-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart