LIF372157412 Life Chemicals Inc.

2-ethoxybenzenesulfonyl chloride

  • MDL number MFCD08706420
  • Cas Number : 68800-33-9
  • Molecular Weight : 220.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart