LIF372157423 Life Chemicals Inc.

{[1-(4-fluorobenzyl)-2,5-dimethyl-1H-pyrrol-3-yl]methyl}amine

  • MDL number MFCD16653336
  • Cas Number : 1177333-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart