LIF372157554 Life Chemicals Inc.

5-(2-Morpholin-4-yl-ethyl)-[1,3,4]thiadiazol-2-ylamine

  • MDL number MFCD07022160
  • Cas Number : 14068-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart