LIF372157559 Life Chemicals Inc.

3-[(Ethylamino)methyl]phenol

  • MDL number MFCD09931765
  • Cas Number : 91239-98-4
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart