LIF372157564 Life Chemicals Inc.

1,3-Benzothiazol-5-amine

  • MDL number MFCD04115282
  • Cas Number : 1123-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart